Learn More
Description
Specifications
Specifications
| Antigen | TRAF-1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulation | Neat whole antisera with 0.02% Sodium Azide |
| Gene Alias | EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1 |
| Gene Symbols | TRAF1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids between 1 and 50 of human TRAF1 was used as immunogen, GenBank no. NP_005649.1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
