Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAF-1 Monoclonal Antibody (1F3), eFluor™ 570, eBioscience™
GREENER_CHOICE

Catalog No. 41979182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-979-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-979-182 Supplier Invitrogen™ Supplier No. 41979182
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibody 1F3 recognizes human TRAF-1. TRAF-1 is a multifunctional adapter molecule and member of the TNF receptor associated factor protein family. TRAF proteins associate with various receptors of the TNF receptor superfamily to mediate signal transduction. TRAF-1 plays a role in regulating cell survival and apoptosis, regulates the signaling of NF kappa B and JNK, and forms a heterodimeric complex with TRAF-2. TRAF-1 also interacts with cell surface receptors such as CD30, CD40, and CD357 (AITR), and cytoplasmic proteins like NIK, RIP2, A20, and TRIP. Expression of TRAF-1 can be induced by Epstein-Barr Virus (EBV) and TNF alpha. Applications Reported: This 1F3 antibody has been reported for use in microscopy, immunohistochemical staining, and immunocytochemistry. Applications Tested: This 1F3 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filter Recommendation: When using this eFluor 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm.

The protein encoded by this gene is a member of the TNF receptor (TNFR) associated factor (TRAF) protein family. TRAF proteins associate with, and mediate the signal transduction from various receptors of the TNFR superfamily. This protein and TRAF2 form a heterodimeric complex, which is required for TNF-alpha-mediated activation of MAPK8/JNK and NF-kappaB. The protein complex formed by this protein and TRAF2 also interacts with inhibitor-of-apoptosis proteins (IAPs), and thus mediates the anti-apoptotic signals from TNF receptors. The expression of this protein can be induced by Epstein-Barr virus (EBV). EBV infection membrane protein 1 (LMP1) is found to interact with this and other TRAF proteins; this interaction is thought to link LMP1-mediated B lymphocyte transformation to the signal transduction from TNFR family receptors. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF-1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone 1F3
Concentration 0.2 mg/mL
Conjugate eFluor 570
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TRAF1
Gene Accession No. Q13077
Gene Alias 4732496E14Rik; EBI6; Epstein-Bar virus-induced protein 6; Epstein-Barr virus-induced protein 6; MGC:10353; TNF receptor associated factor 1; TNF receptor-associated factor 1; Traf1
Gene Symbols TRAF1
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7185
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.