Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-1 Polyclonal Antibody
TRAF-1 Polyclonal Antibody
Supplier: Thermo Scientific PA572916
Description
TRAF-1 Polyclonal Antibody for Western Blot, IHC (P), IP

Specifications
TRAF-1 | |
Polyclonal | |
Unconjugated | |
TRAF1 | |
4732496E14Rik; EBI6; Epstein-Bar virus-induced protein 6; Epstein-Barr virus-induced protein 6; MGC:10353; TNF receptor associated factor 1; TNF receptor-associated factor 1; Traf1 | |
Rabbit | |
50 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P39428 | |
TRAF1 | |
A synthetic peptide corresponding to amino acids 51-69 of mouse TRAF1 was used as immunogen, GenBank no. NP_033447.2. | |
RUO | |
22029 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction