Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-2 Antibody (5C2C3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261661
Description
TRAF-2 Monoclonal specifically detects TRAF-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TRAF-2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 | |
Mouse | |
55.8 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
5C2C3 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
TRAF2 | |
Purified recombinant fragment of human TRAF2 (AA: 39-188) expressed in E. Coli. | |
Protein G purified | |
Apoptosis, Cancer, Cytokine Research, Zinc Finger | |
7186 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction