Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $316.50

Specifications

Antigen TRAF-3
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056175T
SDP
View Documents
Novus Biologicals
NB10056175SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB10056175
SDP
View Documents
Novus Biologicals
NB10056175
0.05 mL
Each for $316.50
Only null left
Add to Cart
 
Description

Description

TRAF-3 Polyclonal specifically detects TRAF-3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

TRAF-3
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Immunology, Innate Immunity, Zinc Finger
Whole antisera with 0.02% Sodium Azide
7187
A synthetic peptide corresponding to amino acis 5-25 (KKMDSPGALQTNPPLKLHTDR) of human TRAF3 was used as immunogen; GenBank no. gi 22027616 ref NP_003291.2 .
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Polyclonal
Antisera
RUO
Human
CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3
TRAF3
IgG
Unpurified
TRAF3
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.