Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-3 Antibody, Novus Biologicals™
SDP

Catalog No. p-5096187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056176T 0.025 mL
NB10056176 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056176T Supplier Novus Biologicals Supplier No. NB10056176SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

TRAF-3 Polyclonal specifically detects TRAF-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen TRAF-3
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:4000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:4000
Formulation Whole antisera with 0.02% Sodium Azide
Gene Alias CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3
Gene Symbols TRAF3
Host Species Rabbit
Immunogen A recombinant protein fragment corresponding to the cytoplasmic end of human TRAF3 (amino acids 341-568) was used as immunogen, GenBank no. GenBank no. gi 22027616 ref NP_003291.2 .
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7187
Test Specificity TRAF3
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.