Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$160.07 - $480.74
Specifications
| Antigen | TRAF-3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1 - 3 ug/ml, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056654T
![]() |
Novus Biologicals
NB10056654SS |
0.05 mL |
Each for $160.07
|
|
|||||
NB10056654
![]() |
Novus Biologicals
NB10056654 |
0.2 mL |
Each for $480.74
|
|
|||||
Description
TRAF-3 Polyclonal specifically detects TRAF-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TRAF-3 | |
| Western Blot 1 - 3 ug/ml, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| NP_663778 | |
| 7187 | |
| A peptide corresponding to amino acids between 300 and 350 of human TRAF3 was used as immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| PBS 0.5%BSA with 0.05% Sodium Azide | |
| CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3 | |
| TRAF3 | |
| IgG | |
| Protein G purified | |
| A 67 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title