Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-4 Antibody (SR1969), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32164850UL
Description
TRAF-4 Monoclonal antibody specifically detects TRAF-4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
TRAF-4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1969 | |
Western Blot 1:1000-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
CART1RING finger protein 83, Cysteine-rich domain associated with RING and Traf domains protein 1, Metastatic lymph node gene 62 protein, MLN 62, MLN62cysteine-rich domain associated with ring and TRAF domain, RNF83malignant 62, TNF receptor-associated factor 4, tumor necrosis receptor-associated factor 4A | |
A synthesized peptide derived from human TRAF-4 (Uniprot #: Q9BUZ4) | |
50 μg | |
Cancer, Zinc Finger | |
9618 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction