Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-5 Antibody (55A219), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001107F
Description
TRAF-5 Monoclonal antibody specifically detects TRAF-5 in Human samples. It is validated for Western BlotSpecifications
TRAF-5 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
55A219 | |
Western Blot | |
MGC:39780, RNF84RING finger protein 84, TNF receptor-associated factor 5 | |
Anti-TRAF5 monoclonal antibody was raised against a fusion protein corresponding to amino acids 77 to 186 of human TRAF5. | |
0.1 mL | |
Apoptosis, Signal Transduction, Zinc Finger | |
7188 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction