Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$243.50 - $353.50
Specifications
| Antigen | TRAF-5 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:2000, Immunohistochemistry, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056178T
![]() |
Novus Biologicals
NB10056178SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB10056178
![]() |
Novus Biologicals
NB10056178 |
0.05 mL |
Each for $353.50
|
|
|||||
Description
TRAF-5 Polyclonal specifically detects TRAF-5 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| TRAF-5 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Signal Transduction, Zinc Finger | |
| Whole antisera with 0.05% Sodium Azide | |
| 7188 | |
| A synthetic peptide corresponding to amino acids 1-20 ([BLAST]MAHSEEQAAVPCAFIRQNSG[/BLAST]) of mouse TRAF5 was used as immunogen, GenBank no. AAH12702.1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Gerbil | |
| MGC:39780, RNF84RING finger protein 84, TNF receptor-associated factor 5 | |
| TRAF5 | |
| IgG | |
| Unpurified | |
| TRAF5 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title