Learn More
TRAF-6 Antibody (326019), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3284MFV610
Description
TRAF-6 Monoclonal antibody specifically detects TRAF-6 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| TRAF-6 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction, Zinc Finger | |
| 7189 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 326019 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6 | |
| E.coli-derived recombinant human TRAF-6, Met1-Val522, Accession # Q9Y4K3 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |