Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177230
Description
TRAF-6 Polyclonal specifically detects TRAF-6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.Specifications
TRAF-6 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6 | |
Rabbit | |
Ion exchange chromatography | |
RUO | |
7189 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 ug/mL | |
NP_004611 | |
TRAF6 | |
Antibody was raised against a peptide corresponding to 14 amino acids near the C-terminus of human TRAF6. The immunogen is located within the last 50 amino acids of TRAF6. Amino Acid Squence: RREGFQPRSTDAGV | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction