Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177230SS
Description
TRAF-6 Polyclonal specifically detects TRAF-6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.Specifications
TRAF-6 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6 | |
Rabbit | |
Ion exchange chromatography | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 ug/mL | |
NP_004611 | |
TRAF6 | |
Antibody was raised against a peptide corresponding to 14 amino acids near the C-terminus of human TRAF6. The immunogen is located within the last 50 amino acids of TRAF6. Amino Acid Squence: RREGFQPRSTDAGV | |
0.025 mg | |
7189 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction