Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TRAF-6 Inhibitor Peptide Set
SDP

Catalog No. NBP226506 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226506 2 mg
NBP2265065 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP226506 Supplier Novus Biologicals™ Supplier No. NBP226506
Add to Cart
Add to Cart

Applications: In vitro assay, Block/Neutralize

Specifications

Host Species Human, Mouse, Rat
Color White
Components TRAF6 Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKRKIPTEDEY (TRAF6 binding sequence: RKIPTEDEY). Molecular weight: 3494. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361.
For Use With (Application) Inhibition of RANKL mediated osteoclast differentiation. Researchers can study the effect of TRAF6 inhibitor peptide using a variety of methods, such as EMSA, NF-kB/p65 ELISA,IkBa phosphorylation ELISA, osteoclast differentiation assay. The Osteoclast formation assay protocol is a guideline which may need to be optimized for different cell types. Use in In vitro assay and blocking neutralizing reported in scientific literature (PMID 24829345)
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 2 mg
Product Type TRAF-6 Inhibitor Peptide Set
Molecular Weight (g/mol) 3494
Inhibitors TRAF-6
Form Lyophilized Solid

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.