Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TRAF-6 Inhibitor Peptide Set
SDP

Catalog No. NBP2265065 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2265065 5 mg
NBP226506 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2265065 Supplier R&D Systems™ Supplier No. NBP2265065MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Applications: In vitro assay, Block/Neutralize

Specifications

Host Species Human, Mouse, Rat
Color White
Components TRAF6 Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKRKIPTEDEY (TRAF6 binding sequence: RKIPTEDEY). Molecular weight: 3494. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361.
For Use With (Application) Blocking/Neutralizing, In vitro assay
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 5 mg
Product Type TRAF-6 Inhibitor Peptide Set
Molecular Weight (g/mol) 3494
Product Line Novus
Inhibitors TRAF-6
Form Lyophilized Solid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.