Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-6 Mouse anti-Human, Alexa Fluor 488, Clone-326019, R&D Systems™

Catalog No. IC3284G025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3284G025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC3284G025 Supplier R&D Systems Supplier No. IC3284G025
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAF-6 Monoclonal specifically detects TRAF-6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen TRAF-6
Applications Flow Cytometry
Classification Monoclonal
Clone 326019
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9Y4K3
Gene Alias E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6, TRAF6
Host Species Mouse
Immunogen E.coli-derived recombinant human TRAF-6 Met1-Val522 Accession # Q9Y4K3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7189
Test Specificity Detects human TRAF-6 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.