Learn More
Invitrogen™ TRAF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587534
Description
Immunogen sequence: MAAASVTPPG SLELLQPGFS KTLLGTKLEA KYLCSACRNV LRRPFQAQCG HRYCSFCLAS ILSSGPQNCA ACVHEGIYEE GISILESSSA FPDNAARREV ESLPAVCPSD GCTWKGTLKE YESCHEGRCP LMLTECPACK GLVRLGEKER HLEHECPERS LSCRHCRAPC CGADVKAHHE VCPKFPLTCD GCGKKKIPRE KFQDHVKTCG KCRVPCRFHA IGCLETVEGE KQQEHEVQWL REHLAMLLSS VLEAKPLLGD QSHAGSELLQ RCESLEKKTA TFENIVCVLN REVERVAMTA; Positive Samples: HeLa, 293T, THP-1, PC-12, Ramos, Jurkat, NIH/3T3; Cellular Location: Cytoplasm.
Tumor necrosis factor (TNF) receptor associated factors (TRAFs) were initially discovered as adaptor proteins that link the TNF receptor superfamily to signaling pathways and are thus important regulators of cell death and cellular response to stress. TRAF proteins share a homology region that allows them to bind to cell receptors and other TRAF proteins, causing the activation of different signal cascades depending on the TRAFs involved. For example, TRAF2 and TRAF3 directly bind to the CD40, a NF receptor superfamily member involved in inducing B cell immunity, and are critical for NF-kappa-B activation in mouse B lymphocytes. TRAF2 along with TRAF6 has also been shown to be required for CD40 signaling in nonhemopoietic cells. TRAF2 also interacts with the TRFR superfamily member lymphotoxin-beta receptor (LTbetaR) in association with TRAF3 and the apoptosis inhibitors cIAP1 and Smac.
Specifications
TRAF2 | |
Polyclonal | |
Unconjugated | |
TRAF2 | |
AI325259; E3 ubiquitin-protein ligase TRAF2; MGC:45012; OTTHUMP00000064745; RING-type E3 ubiquitin transferase TRAF2; TNF receptor associated factor 2; Tnf receptor-associated factor 2; TRAF2; TRAP; TRAP3; tumor necrosis factor type 2 receptor associated protein 3; tumor necrosis factor type 2 receptor-associated protein 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22030, 311786, 7186 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2.71 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P39429, Q12933 | |
TRAF2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TRAF2 (NP_0669612). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.