Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735475 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5475 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5475 Supplier Proteintech Group Inc Supplier No. 268461AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tumor necrosis factor (TNF) induced signaling is mediated through association of TNF receptor (TNFR) with adaptor proteins, such as TNF receptor associated proteins (TRAFs). TRAFs form a family of cytoplasmic adapter proteins that mediate signal transduction from many members of the TNF-receptor superfamily (e.g. RANK, CD30, CD40, etc.) and the interleukin-1 receptor. The carboxy-terminal region of TRAFs is required for self-association and interaction with receptor cytoplasmic domains following ligand-induced oligomerization. Recent molecular cloning studies have lead to identification of six TRAFs (TRAF1-TRAF6) (1-4). TRAF2 is a 502-amino acid protein. Mutagenic studies suggest that the N-terminal RING finger and two adjacent zinc fingers of TRAF2 are required for NF-kB activation, where as interaction with TNFR is mediated through C-terminus domain. Distinct domains in the N- and -C termini are also required for association with TRAF1 and protein kinase receptor interacting protein (RIP). TRAF2 is involved in cellular resistant to TNF-induced apoptosis. TRAF-2 deficient mice appeared normal at birth, however, they die prematurely, probably due to atrophy of thymus, spleen, muscle mass and lack of TRAF-2′s cytoprotective role.

Specifications

Antigen TRAF2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TRAF2
Gene Accession No. Q12933
Gene Alias TRAF2, TRAP, TRAP3
Gene Symbols TRAF2
Host Species Rabbit
Immunogen TRAF2 Fusion Protein Ag25387
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7186
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.