Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA587516
Description
Immunogen sequence: MESSKKMDSP GALQTNPPLK LHTDRSAGTP VFVPEQGGYK EKFVKTVEDK YKCEKCHLVL CSPKQTECGH RFCESCMAAL LSSSSPKCTA CQESIVKDKV; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cytoplasm, Endosome, Mitochondrion
Tumor necrosis factor (TNF) induced signaling is mediated through association of TNF receptor (TNFR) with adaptor proteins, such as TNF receptor associated proteins (TRAFs). TRAFs form a family of cytoplasmic adapter proteins that mediate signal transduction from many members of the TNF-receptor superfamily (e. g. RANK, CD30, CD40, etc. ) and the interleukin-1 receptor. The carboxy-terminal region of TRAFs is required for self-association and interaction with receptor cytoplasmic domains following ligand-induced oligomerization. Recent molecular cloning studies have lead to identification of six TRAFs (TRAF1-TRAF6). TRAF3, originally named CRAF1, interacts directly with the CD40 cytoplasmic tail through a region of similarity to the tumor necrosis factor-alpha (TNF-alpha) receptor-associated factors. TRAF3 binds only a single site, which contains the sequence PEQET, whereas TRAF1 and TRAF2 are capable of binding to either the PEQET site or an additional downstream domain.
Specifications
TRAF3 | |
Polyclonal | |
Unconjugated | |
TRAF3 | |
AI528849, amn, CAP1, CAP-1, CD40 associated protein 1, CD40 binding protein, CD40 receptor associated factor 1, CD40 receptor-associated factor 1, CD40-binding protein, CD40BP, CRAF1, IIAE5, LAP1, LMP1-associated protein 1, RING-type E3 ubiquitin transferase TRAF3, T-BAM, TNF receptor associated factor 3, Tnf receptor-associated factor 3, TRAF3, Trafamn | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7187 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
Q13114 | |
TRAF3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TRAF3 (NP_0032912) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction