Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TRAF3, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE861 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE861 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE861 Supplier MilliporeSigma™ Supplier No. ABE861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TNF receptor-associated factor 3 (UniProt Q13114; also known as CAP-1, CD40 receptor-associated factor 1, CD40-binding protein, CD40BP, CRAF1, LAP1, LMP1-associated protein 1) is encoded by the TRAF3 (also known as CAP1, CRAF1) gene (Gene ID 7187) in human. NF-κB-inducing kinase (NIK) is negatively regulated by a complex composed of the E3 ubiquitin ligases c-IAP1 and 2 (cellular inhibitor of apoptosis 1 and 2) and the adaptors TRAF2 and 3 (TNF receptor-associated factors 2 and 3). The TRAF2-TRAF3 complex bridges the c-IAPs and NIK into close proximity, where TRAF2 recruits c-IAP1/2 and TRAF3 binds to NIK. NF-κB signaling activation results in TRAF3 degradation, leading to upregulated NIK. TRAF3 deletion in B-cells results in uncontrolled growth and TRAF3 mutations have been identified in several human B-cell cancers, including Hodgkin lymphoma, splenic marginal zone lymphoma, and multiple myeloma. TRAF3 also plays an indirect role in promoting PI3K/AKT and JAK/STAT signalings, which are negatively regulated by NIK. TRAF3 prevents NIK from targeting PTEN to the plasma membrane and thereby promoting PI3K/AKT signaling. TRAF3 is required for normal T cells to function properly and anaplastic large cell lymphoma (ALCL) cells reply on PI3K/AKT and JAK/STAT signaling for proliferation. Opposite to B-cell cancers, TRAF3 knockdown triggers cell cycle arrest in ALCL cells. Human TRAF3 contains one RING-type (a.a. 68-77) and two TRAF-type (a.a. 135-190 and 191-249) zinc fingers, a , coiled coil (a.a. 267-338) regin, followed a C-terminal MATH (meprin and TRAF homology) domain (a.a. 415-560) that mediates interaction with receptor cytomplasmic domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF3
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene TRAF3, CAP1, CRAF1
Gene Accession No. Q13114
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence within the coiled coil region of human TRAF3
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003291
Test Specificity This rabbit polyclonal antibody targets a coiled coil region sequence conserved among bovine, equine, human, mouse, and rat TRAF3 protein sequences.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.