Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRAF3IP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530507
Description
Recommended positive controls: U87-MG, mouse testis, rat testis. Predicted reactivity: Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Tumor necrosis factor receptor (TNFR) superfamily members transmit signals regulating proliferation, differentiation and apoptosis in various types of cells. TNFR-associated factors (TRAFs) are a family of proteins that were initially discovered as downstream signal transducers of the TNFR superfamily. TRAF3 contains an N-terminal ring finger/zinc finger region that is thought to be essential for downstream signaling. MIP-T3 is associated with TRAF3. MIP-T3 binds to taxol-stabilized microtubules and to tubulin in vitro, and MIP-T3 recruits TRAF3 to microtubules when both proteins are overexpressed. The MIP-T3/TRAF3 interaction requires the coiled-coil TRAF-N domain of TRAF3. This interaction may provide a novel mechanism in sequestering TRAF3 to the cytoskeletal network.
Specifications
| TRAF3IP1 | |
| Polyclonal | |
| Unconjugated | |
| TRAF3IP1 | |
| 3930402D05Rik; AU041749; IFT54; interleukin 13 receptor alpha 1-binding protein-1; interleukin-13 receptor alpha 1-binding protein 1; Intraflagellar transport protein 54 homolog; microtubule interacting protein that associates with TRAF3; microtubule-interacting protein associated with TRAF3; microtubule-interacting protein that associates with TRAF3; MIPT3; MIP-T3; SLSN9; TNF receptor-associated factor 3 interacting protein 1; TRAF3 interacting protein 1; TRAF3-interacting protein 1; TRAF3IP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26146, 363286, 74019 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q149C2, Q5XIN3, Q8TDR0 | |
| TRAF3IP1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human TRAF3IP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction