Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAF3IP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550277
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550277 Supplier Invitrogen™ Supplier No. PA550277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total TRAF3IP1 protein.

Tumor necrosis factor receptor (TNFR) superfamily members transmit signals regulating proliferation, differentiation and apoptosis in various types of cells. TNFR-associated factors (TRAFs) are a family of proteins that were initially discovered as downstream signal transducers of the TNFR superfamily. TRAF3 contains an N-terminal ring finger/zinc finger region that is thought to be essential for downstream signaling. MIP-T3 is associated with TRAF3. MIP-T3 binds to taxol-stabilized microtubules and to tubulin in vitro, and MIP-T3 recruits TRAF3 to microtubules when both proteins are overexpressed. The MIP-T3/TRAF3 interaction requires the coiled-coil TRAF-N domain of TRAF3. This interaction may provide a novel mechanism in sequestering TRAF3 to the cytoskeletal network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF3IP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TRAF3IP1
Gene Accession No. Q149C2, Q8TDR0
Gene Alias 3930402D05Rik; AU041749; IFT54; interleukin 13 receptor alpha 1-binding protein-1; interleukin-13 receptor alpha 1-binding protein 1; Intraflagellar transport protein 54 homolog; microtubule interacting protein that associates with TRAF3; microtubule-interacting protein associated with TRAF3; microtubule-interacting protein that associates with TRAF3; MIPT3; MIP-T3; SLSN9; TNF receptor-associated factor 3 interacting protein 1; TRAF3 interacting protein 1; TRAF3-interacting protein 1; TRAF3IP1
Gene Symbols TRAF3IP1
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human TNF receptor-associated factor 3 interacting protein 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26146, 74019
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.