Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF3IP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 11 publications
$207.00 - $501.50
Specifications
| Antigen | TRAF3IP2 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Simple Western 1:20, Immunohistochemistry, Immunoprecipitation reported in scientific literature (PMID 24561578), Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056740T
![]() |
Novus Biologicals
NB10056740SS |
0.025 mg |
Each for $207.00
|
|
|||||
NB10056740
![]() |
Novus Biologicals
NB10056740 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
TRAF3IP2 Polyclonal specifically detects TRAF3IP2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| TRAF3IP2 | |
| Western Blot 1-2 ug/ml, Simple Western 1:20, Immunohistochemistry, Immunoprecipitation reported in scientific literature (PMID 24561578), Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| NP_679211 | |
| 10758 | |
| This antibody was developed against a synthetic peptide (QDLPRPLRSREFPQFEP) corresponding to amino acids 225-241 of human TRAF3IP2. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/ml | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| ACT1DKFZP586G0522, adapter protein CIKS, C6orf2, C6orf4chromosome 6 open reading frame 5, C6orf5DKFZp586G0522, C6orf6MGC3581, CIKSchromosome 6 open reading frame 2, Connection to IKK and SAPK/JNK, NFkB-activating protein ACT1, Nuclear factor NF-kappa-B activator 1, TRAF3 interacting protein 2, TRAF3-interacting protein 2 | |
| TRAF3IP2 | |
| IgG | |
| Protein G purified | |
| An approx. 60 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title