Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF3IP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519989
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519989 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519989 Supplier Thermo Scientific Supplier No. PA519989
Add to Cart
Add to Cart

TRAF3IP2 Polyclonal Antibody

TRAF3IP2 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF3IP2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TRAF3IP2
Gene Accession No. O43734
Gene Alias ACT1; adapter protein CIKS; AI429613; C6orf2; C6orf4; C6orf5; C6orf6; CANDF8; CIKS; connection to IKK and SAPK/JNK; NFkB-activating protein ACT1; nuclear factor NF-kappa-B activator 1; PSORS13; RGD1304768; RP3-487J7.3; TRAF3 interacting protein 2; TRAF3-interacting protein 2; TRAF3IP2
Gene Symbols TRAF3IP2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 2 to 15 of human CIKS.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10758
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.