Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAF3IP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106565
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106565 Supplier Invitrogen™ Supplier No. PA5106565
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total T3JAM.

The gene encodes a protein that mediates cell growth by modulating the c-Jun N-terminal kinase signal transduction pathway. The encoded protein may also interact with a large multi-protein assembly containing the phosphatase 2A catalytic subunit. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF3IP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRAF3IP3
Gene Accession No. Q8C0G2, Q9Y228
Gene Alias 6030423D04Rik; AI115021; DJ434O14.3; FLJ44151; MGC117354; MGC163289; RGD1304848; T3JAM; TRAF3 interacting Jun N terminal kinase (JNK) activating modulator; TRAF3 interacting protein 3; TRAF3-interacting JNK-activating modulator; TRAF3-interacting JNK-activating modulator-like protein; TRAF3-interacting Jun N-terminal kinase (JNK)-activating modulator; TRAF3-interacting protein 3; Traf3ip3
Gene Symbols TRAF3IP3
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRAF3IP3(Accession Q9Y228), corresponding to amino acid residues C256-R306.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215243, 360900, 80342
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.