Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547473
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547473 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547473 Supplier Invitrogen™ Supplier No. PA547473
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, less than 1% cross-reactivity with recombinant human TRAF-1, -2, -3, -5, or -6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The tumor necrosis factor (TNF) receptor superfamily is composed of several type I integral membrane glycoproteins that exhibit homology in their cystine rich extracellular domains. Members of this family include TNF-RI, TNF-RII and CD40. Ligands for these receptors can be small, secreted proteins, such as TNF, or type II integral membrane proteins, as is the case for the CD40 ligand, CD40L. While the signal transduction mechanism of the TNF receptor superfamily is poorly understood, activation of TNF-R or CD40 have been shown to induce the nuclear translocation of NFkB. Members of the TRAF (TNF receptor-associated factor) family have been implicated in this process. Four members have thus far been described and are designated TRAF1, TRAF2, TRAF3 (variously referred to as CRAF1, LAP1 or CD40bp) and TRAF4. TRAF4, originally termed CART1, is specifically expressed in breast carcinomas, and is localized to the nucleus in such tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF4
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TRAF4
Gene Accession No. Q9BUZ4
Gene Alias A530032M13Rik; Cart1; cysteine-rich domain associated with ring and TRAF domain; cysteine-rich domain associated with RING and Traf domains protein 1; Cysteine-rich motif associated to RING and Traf domains protein 1; malignant 62; metastatic lymph node gene 62 protein; MLN 62; MLN62; msp2; RING finger protein 83; RNF83; RNF83malignant 62; TNF receptor associated factor 4; TNF receptor-associated factor 4; tnf receptor-associated factor 4b; traf4; TRAF4 protein; TRAF4 variant 6; traf4b; tumor necrosis receptor-associated factor 4A
Gene Symbols TRAF4
Host Species Goat
Immunogen E. coli-derived recombinant human TRAF-4 Met1-Ser470.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9618
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.