Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527705 Supplier Invitrogen™ Supplier No. PA527705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, Jurkat. Predicted reactivity: Mouse (84%), Dog (91%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Tumor necrosis factor (TNF) induced signaling is mediated through association of TNF receptor (TNFR) with adaptor proteins, such as TNF receptor associated proteins (TRAFs). TRAFs form a family of cytoplasmic adapter proteins that mediate signal transduction from many members of the TNF-receptor superfamily (e. g. RANK, CD30, CD40, etc. ) and the interleukin-1 receptor. The carboxy-terminal region of TRAFs is required for self-association and interaction with receptor cytoplasmic domains following ligand-induced oligomerization. Recent molecular cloning studies have lead to identification of six TRAFs (TRAF1-TRAF6) (1-4). TRAF5 is a 558-amino acid protein. TRAF5 is implicated in NF-kB and c-Jun NH(2)-terminal kinase/stress-activated protein kinase activation by members of the TNF receptor superfamily, including CD27, CD30, CD40, and lymphotoxin-beta receptor. Targeted disruption of TRAF5 gene causes defects in CD40-CD27 mediated lymphocyte activation (5).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene TRAF5
Gene Accession No. O00463
Gene Alias 2700049M22Rik; AI875481; AU022477; BH3 interacting domain death agonist; BH3-interacting domain death agonist; BH3-interacting domain death agonist p11; BH3-interacting domain death agonist p13; BH3-interacting domain death agonist p15; BID; MGC:39780; p11 BID; p13 BID; p15 BID; p22 BID; RING finger protein 84; RNF84; TNF receptor associated factor 5; TNF receptor-associated factor 5; Traf5
Gene Symbols TRAF5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human TRAF5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7188
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.