Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TRAF7 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7449
![]() ![]() |
Proteintech Group Inc
117801AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TRAFs are signal transducers for members of TNFR superfamily. These are critically involved in activation of MAP3K family members involved in NF-kappaB and JNK activation and apoptosis. TRAF7, a novel 670 A. A protein, is a human homolog of mouse Traf7 andconsists of an N-terminal RING finger domain, central TRAF domain, a coiled-coil motif and 7 WD40 repeats at the C-terminus. It has E3-ubiquitin ligase activity for self-ubiquitination. TRAF7 specifically interacts with MEKK3 and mediates MEKK3-induced AP1 and CHOP activation and induces apoptosis through Caspase-independent pathways. It is a multifunctional protein involved in sumoylation and negative regulation of c-Myb, thereby regulating proliferation and differentiation of hematopoietic cells.Specifications
TRAF7 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q6Q0C0, Q922B6 | |
224619, 360491, 84231 | |
TRAF7 Fusion Protein Ag2414 | |
Primary | |
-20°C | |
TRAF7 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
RFWD1, RING finger protein 119, RNF119, TRAF7 | |
Traf7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title