Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAFD1 Monoclonal Antibody (8E6E7)
GREENER_CHOICE

Catalog No. PIMA517190
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517190 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517190 Supplier Invitrogen™ Supplier No. MA517190
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17190 targets TRAFD1 in indirect ELISA, FACS, ICC, IF, and WB applications and shows reactivity with Human samples. The MA5-17190 immunogen is purified recombinant fragment of human TRAFD1 (amino acids: 401-582) expressed in E. Coli. MA5-17190 detects TRAFD1 which has a predicted molecular weight of approximately 64.8kDa.

The innate immune system confers host defense against viral and microbial infection, and TRAFD1 is a negative feedback regulator that controls excessive immune responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAFD1
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8E6E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene TRAFD1
Gene Accession No. O14545
Gene Alias 1110008K06Rik; Fln29; FLN29 gene product; Protein FLN29; TRAF type zinc finger domain containing 1; TRAFD1; TRAF-type zinc finger domain containing 1; TRAF-type zinc finger domain-containing protein 1
Gene Symbols TRAFD1
Host Species Mouse
Immunogen Purified recombinant fragment of human TRAFD1 (amino acids: 401-582) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.