Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAFD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540603 Supplier Invitrogen™ Supplier No. PA540603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TATNHVTEGI PRLDSQPQET SPELPRRRVR HQGDLSSGYL DDTKQETANG Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 85%; Rabbit: 86%; Rat: 86%.

The innate immune system confers host defense against viral and microbial infection, and TRAFD1 is a negative feedback regulator that controls excessive immune responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAFD1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRAFD1
Gene Accession No. O14545
Gene Alias 1110008K06Rik; Fln29; FLN29 gene product; Protein FLN29; TRAF type zinc finger domain containing 1; TRAFD1; TRAF-type zinc finger domain containing 1; TRAF-type zinc finger domain-containing protein 1
Gene Symbols TRAFD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TRAFD1 (aa 395-444).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10906
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.