Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAFD1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA558712
Description
Immunogen sequence: PSSPCVPKLS NSDSQDIQGR NRDSQNGAIA PGHVSVIRPP QNLYPENIVP SFSPGPSGRY GASGRSEGGR NSRVTPAAAN YRSRTAKAKP SKQQGAGD Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 67%.
The innate immune system confers host defense against viral and microbial infection, and TRAFD1 is a negative feedback regulator that controls excessive immune responses
Specifications
TRAFD1 | |
Polyclonal | |
Unconjugated | |
TRAFD1 | |
TRAF-type Zinc finger domain containing 1, FLN29 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10906 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14545 | |
TRAFD1 | |
Recombit protein corresponding to Human TRAFD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction