Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform™ Traffic Sign
Shop All AccuformNMC Products
Click to view available options
Displayed Text:
BE PREPARED TO STOP
EMERGENCY MEDICAL CARE
H (Hospital Symbol)
Description
- Durable, .080 in. aluminum signs will not rust
- Engineer-Grade Prismatic Reflective sign face provides visibility in low-light conditions
- UV-resistant inks prevent fading from long-term outdoor use
- Clear UV Protective Overcoat
- Post mount holes
- Meets ASTM D 4956 Type I
Specifications
Specifications
| Certifications/Compliance | ASTM D 4956 Type I |
| Grade | Engineer |
| Material | Reflective Aluminum |
| Mounting Type | Corner Holes |
| Languages | English |
| Dimensions (L x W) | 30 x 30 in. |
| Displayed Text | BE PREPARED TO STOP |
| Color | Black on Yellow |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction