Learn More
Invitrogen™ TRAG3 Polyclonal Antibody

Description
This protein is expressed in various tumor cell lines including carcinomas, myeloid or lymphoid malignancies, melanomas or prostate cancer. It is also weakly expressed in kidney and overexpressed in taxol-resistant breast cancer line MDA 435(TR) or the doxorubicin-resistant multiple myelanoma lines 8226/Dox(40) or 8226/MDR(10)V.1.
Specifications
Specifications
| Antigen | TRAG3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | CSAG2 |
| Gene Accession No. | Q9Y5P2 |
| Gene Alias | cancer/testis antigen 24.2; cancer/testis antigen family 24, member 2; chondrosarcoma-associated gene 2/3 protein; chondrosarcoma-associated gene 2/3 protein-like; CSAG family member 2; CSAG family, member 3B; CSAG2; CSAG3B; CT24.2; MGC149851; MGC149852; taxol resistance associated gene 3; taxol-resistant-associated gene 3 protein; trag 3; TRAG3; TRAG-3 |
| Gene Symbols | CSAG2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.