Learn More
Tragacanth powder
Description
Tragacanth powder is used as an emulsifier, thickener, stabilizer in food and pharmaceutical industry. It acts as a binder used in the preparation of artist's pastels. It is used as a natural hair shampoo. It plays an important role as freezing-embedding medium for tissue preservation. It is used as a demulcent in cough and cold preparations as well as to manage diarrhea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Merck Index | 14,4581 |
| Solubility Information | Slightly soluble in water. |
| Quantity | 100 g |
| Odor | Odorless |
| Chemical Name or Material | Tragacanth powder |
Safety and Handling
| EINECSNumber | 232-552-5 |
| RTECSNumber | XW7750000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.