Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRAIL R2/TNFRSF10B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6313SP
This item is not returnable.
View return policy
Description
TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Western Blot.Specifications
TRAILR2/TNFRSF10B | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
NP_003833 | |
TNFRSF10B | |
E. coli-derived recombinant human TRAIL R2/TNFRSF10B Ile56-Pro128 Accession # NP_003833 | |
25 μg | |
Primary | |
Detects human TRAIL R2/TNFRSF10B in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhOPG, rhRANK, rhRELT, rhTNF RI, rhTNF RII, rhTRAIL R1, rhTRAIL R3, rhTRAIL R4, or rhTWEAK R is observed. Additionally in direct ELISAs, no cross-reactivity with rhTRAIL R4, rhXEDAR, recombinant mouse (rm) DR5, rhTRAIL R1, rmTRAIL R2, or rmTNFRH3 is observed. | |
Human | |
Purified |
Western Blot | |
603307 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TNFRSF10B, TRAIL receptor 2, TRAILR2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8795 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction