Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAIL-R4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596515
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596515 Supplier Invitrogen™ Supplier No. PA596515
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLFRRRSCPS RVPGAEDNAR NETLSNRYLQ PTQVSEQEIQ GQELAELTGV TVELPEEPQR LLEQAEAEGC QRRRLLVPVN DADSADISTL LDASATLEEG HAKETIQDQL VGSEKLFYEE DEAGSATSCL; Positive Samples: A-549, 293T; Cellular Location: Membrane, Single-pass type I membrane protein.

TRAIL (also known as Apo-2L) is a member of the tumor necrosis factor (TNF) ligand family that rapidly induces apoptosis in a variety of transformed cell lines. A fourth member of the emerging TRAIL receptor family, TRAIL-R4, has signaling capacity similar to that of TRAIL-R1 and TRAIL-R2 with respect to NF-kappaB activation, but differs in its inability to induce apoptosis. Like TRAIL-R1 and TRAIL-R2, this receptor shows widespread tissue expression. The human TRAIL-R4 gene has been mapped to chromosome 8p22-21, clustered with three other TRAIL receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAIL-R4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TNFRSF10D
Gene Accession No. Q9UBN6
Gene Alias CD264; DCR2; Decoy receptor 2; TNF receptor superfamily member 10d; TNF receptor-related receptor for TRAIL; TNF-related apoptosis-inducing ligand receptor 4; TNFRSF10D; TRAIL receptor 4; TRAIL receptor with a truncated death domain; TRAILR4; TRAIL-R4; TRUNDD; Tumor necrosis factor receptor superfamily member 10D; tumor necrosis factor receptor superfamily member 10d, decoy with truncated death domain; tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain; UNQ251/PRO288
Gene Symbols TNFRSF10D
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 257-386 of human TNFRSF10D (NP_0038312).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8793
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.