Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAIL/TNFSF10 Antibody (55B709.3) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 8 publications

$206.50 - $499.50

Specifications

Antigen TRAIL/TNFSF10
Clone 55B709.3
Concentration 1.0 mg/mL
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056518T
SDP
View Documents
Novus Biologicals
NB10056518SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056518
SDP
View Documents
Novus Biologicals
NB10056518
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

TRAIL/TNFSF10 Monoclonal specifically detects TRAIL/TNFSF10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TRAIL/TNFSF10
1.0 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Signal Transduction, Tumor Suppressors
NP_003801.1, P50592
8743
This monoclonal antibody was raised against a peptide corresponding to amino acids 17-35 of human TRAIL. This peptide sequence has been shown to be involved in TRAIL and DR5 (TRAIL-R2) interaction.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
55B709.3
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Monoclonal
Purified
RUO
Human
Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
TNFSF10
IgG1 κ
Protein G purified
In Jurkat, a 32 kDa band, which represents a membrane bound form of TRAIL, is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.