Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAIL/TNFSF10 Antibody (B-S23), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314602APC
Description
TRAIL/TNFSF10 Monoclonal antibody specifically detects TRAIL/TNFSF10 in Human samples. It is validated for Flow Cytometry, ELISA, Functional AssaySpecifications
TRAIL/TNFSF10 | |
Monoclonal | |
APC | |
Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10 | |
Recombinant human TRAIL/TNFSF10 | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Signal Transduction, Tumor Suppressors | |
8743 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA, Functional Assay | |
B-S23 | |
PBS | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction