Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (001), Alexa Fluor™ 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289475AF405
Description
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
TRAILR1/TNFRSF10A | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human TNFRSF10B Human TNFRSF10D | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
1 | |
ELISA, Sandwich ELISA Detection | |
APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAILR1/TNFRSF10A (NP_003835.2; Met1-Asn239). | |
0.1 mL | |
Apoptosis, Cancer | |
8797 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction