Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (32A1380) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 11 publications
Supplier: Novus Biologicals NBP227350
Description
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready, Block/Neutralize.Specifications
TRAILR1/TNFRSF10A | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 2 - 4 ug/mL, Simple Western, Flow Cytometry 1 uL/1 million cells, Flow (Cell Surface) reported in scientific literature (PMID 18004565), Flow (Intracellular) reported in scientific literature (PMID 17541421), CyTOF-ready, Block/Neutralize | |
APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Blocking Assay, Flow Cytometry, CyTOF | |
32A1380 | |
Unconjugated | |
O00220 | |
TNFRSF10A | |
This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4. | |
0.1 mg | |
Apoptosis, Cancer | |
8797 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction