Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR1/TNFRSF10A Antibody (32A1380) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 22 publications

$206.50 - $499.50

Specifications

Antigen TRAILR1/TNFRSF10A
Clone 32A1380
Concentration 1.0 mg/mL
Dilution Western Blot 2-4 ug/ml, Simple Western 20 ug/ml, Flow Cytometry 1:20-1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 26044191), Flow (Cell Surface) reported in scientific literature (PMID 18004565), Flow (Intracellular) reported in scientific literature (PMID 17541421), CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056747T
SDP
View Documents
Novus Biologicals
NB10056747SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056747
SDP
View Documents
Novus Biologicals
NB10056747
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready.
Specifications

Specifications

TRAILR1/TNFRSF10A
1.0 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
Unconjugated
Mouse
Apoptosis, Cancer
O00220
8797
This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
32A1380
Western Blot 2-4 ug/ml, Simple Western 20 ug/ml, Flow Cytometry 1:20-1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 26044191), Flow (Cell Surface) reported in scientific literature (PMID 18004565), Flow (Intracellular) reported in scientific literature (PMID 17541421), CyTOF-ready
Monoclonal
Purified
RUO
Human
APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a
TNFRSF10A
IgG1 κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.