Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (32A242), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056528F
Description
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TRAILR1/TNFRSF10A | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TNFRSF10A | |
This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4. | |
0.1 mL | |
Apoptosis, Cancer | |
8797 | |
Human, Mouse, Rat | |
Purified |
Immunocytochemistry, Immunofluorescence | |
32A242 | |
Immunocytochemistry/Immunofluorescence | |
APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
In HeLa, a 57 kDa band should be observed. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction