Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (DR-4-02), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP231343AF647
Description
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
TRAILR1/TNFRSF10A | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TNFRSF10A | |
Fusion protein (Human). Fusion protein containing the extracellular part of DR4 and the constant part of the heavy chain of the human IgG1 (NM_003844.3). | |
0.1 mL | |
Apoptosis, Cancer | |
8797 | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
DR-4-02 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal antibody DR-4-02 recognizes an extracellular epitope of TRAIL-R1 (DR4), a human death receptor 4 expressed in most human tissues (spleen, peripheral blood leucocytes, thymus) and in a variety of tumour-derived cell lines. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction