Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (TR1.02), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280069IR
This item is not returnable.
View return policy
Description
TRAILR1/TNFRSF10A Monoclonal antibody specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TRAILR1/TNFRSF10A | |
Monoclonal | |
DyLight 755 | |
APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
Recombinant human TRAILR1/TNFRSF10A (DR4). | |
0.1 mL | |
Apoptosis, Cancer | |
8797 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
TR1.02 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction