Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR2/TNFRSF10B Antibody (102), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289513UV
Description
TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
TRAILR2/TNFRSF10B | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human TNFRSF10A/ CD261/ APO2 Human TNFRSF10D/ CD264/ DCR2/ TRAILR4 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, ELISA | |
102 | |
Flow Cytometry, ELISA | |
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAILR2/TNFRSF10B extracellular domain (Met 1-Glu 182; NP_003833.3). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway | |
8795 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction