Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR2/TNFRSF10B Antibody (131), DyLight 550

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290575R
Description
TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Mouse samples. It is validated for ELISA.Specifications
TRAILR2/TNFRSF10B | |
Monoclonal | |
DyLight 550 | |
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TRAILR2/TNFRSF10B (NP_064671.2; Met 1-Ser 177). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway | |
8795 | |
Mouse | |
Purified |
ELISA | |
131 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human TNFRSF10B | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction