Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR2/TNFRSF10B Antibody - Azide Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 19 publications
Supplier: Novus Biologicals NB10055744
Description
TRAILR2/TNFRSF10B Polyclonal specifically detects TRAILR2/TNFRSF10B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy, Flow (Cell Surface).Specifications
TRAILR2/TNFRSF10B | |
Polyclonal | |
Unconjugated | |
PBS with No Preservative | |
TNFRSF10B | |
Rabbit anti-DR5 polyclonal antibody was raised against a peptide corresponding to amino acids 388 to 407 (isoform 2) of human DR5 precursor (1,2). The same sequence is found in isoform 1 at amino acids 417-436. | |
0.1 mg | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
0.5mg/mL | |
Western Blot 5 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy reported in scientific literature (PMID 16426839), Flow (Cell Surface) reported in scientific literature (PMID 16426839) | |
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
Rabbit | |
Protein G purified | |
RUO | |
8795 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction