Learn More
Specifications
Specifications
| Gene ID (Entrez) | 8795 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-55744 |
| Gene Alias | CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2 L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2 B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10 B, tumor necrosis factor receptor superfamily, member 10 b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 |
| Gene Symbol | TNFRSF10B |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
