Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TRAILR2/TNFRSF10B Antibody Blocking Peptide
SDP

Catalog No. NB398689 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398689 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398689 Supplier Novus Biologicals™ Supplier No. NB10056618PEP
Add to Cart
Add to Cart

Specifications

Gene ID (Entrez) 8795
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-56618
Gene Alias CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2 L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2 B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10 B, tumor necrosis factor receptor superfamily, member 10 b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Gene Symbol TNFRSF10B
Quantity 0.1 mg
Research Category Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.