Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR2/TNFRSF10B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.00 - $401.00
Specifications
Antigen | TRAILR2/TNFRSF10B |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TRAILR2/TNFRSF10B Polyclonal specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Western Blot.Specifications
TRAILR2/TNFRSF10B | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
TNFRSF10B | |
IgG | |
Affinity Purified |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer | |
PBS with 0.02% Sodium Azide | |
8795 | |
Reacts with residues 280-296 (EHLLEPAEAERSQRRRL) of the human TRAIL-R2/DR5 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title